Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EMID1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | EMID1 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
EMID1 Polyclonal specifically detects EMID1 in Human samples. It is validated for Western Blot.Specifications
EMID1 | |
Polyclonal | |
Rabbit | |
Q96A84 | |
129080 | |
Synthetic peptides corresponding to EMID1(EMI domain containing 1) The peptide sequence was selected from the C terminal of EMID1. Peptide sequence TMIGLYEPELGSGAGPAGTGTPSLLRGKRGGHATNYRIVAPRSRDERG. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
EMI domain containing 1, EMI5, Emilin and multimerin domain-containing protein 1, emilin and multimerin-domain containing protein 1, Emu1, EMU1EMI domain-containing protein 1, hEmu1, MGC50657, putative emu1 | |
EMID1 | |
IgG | |
45 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title