Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EMID2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $646.00
Specifications
Antigen | EMID2 |
---|---|
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Host Species | Rabbit |
Description
EMID2 Polyclonal specifically detects EMID2 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
EMID2 | |
Unconjugated | |
Rabbit | |
Human | |
136227 | |
This antibody was developed against a recombinant protein corresponding to the amino acid sequence:ANLVSYRTLIRPTYRVSYRTVTVLEWRCCPGFTGSNCDEECMNCTRLSDMSERLTTLEAKVLLLEAAERPSSPDNDLPAPESTPPTWNEDFLPDAIPLAHPVP | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Polyclonal | |
Purified | |
RUO | |
COL26A1, EMI domain containing 2, EMI domain-containing protein 2, Emilin and multimerin domain-containing protein 2, emilin and multimerin-domain containing protein 2, EMU2, Emu2type XXVI, alpha 1, hEmu2, KIAA1299, MGC129848, putative emu2, SH2B, SH2B1 | |
EMID2 | |
IgG | |
Protein A purified |
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title