Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EMILIN1 Antibody (C11A8) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB019672 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.02 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB019672 0.02 mg
NBP236338 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB019672 Supplier Novus Biologicals Supplier No. NBP2363380.02MG
Add to Cart
Add to Cart

Rat Monoclonal Antibody

EMILIN1 Monoclonal specifically detects EMILIN1 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.

Specifications

Antigen EMILIN1
Applications Immunofluorescence, Immunohistochemistry (Frozen), Immunocytochemistry, Western Blot, Immunohistochemistry
Classification Monoclonal
Clone C11A8
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 2-10ug/ml, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
Formulation PBS with No Preservative
Gene Alias DKFZp586M121, elastin microfibril interface-located protein 1, elastin microfibril interfacer 1, EMI, EMILIN, EMILIN-1, gp115
Gene Symbols EMILIN1
Host Species Rat
Immunogen recombinant mouse Emilin-1
Purification Method Protein G purified
Quantity 0.02 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11117
Reconstitution Centrifuge vial prior to opening. Reconstitute in sterile water to a concentration of 0.1-1.0 mg/ml.
Target Species Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.