Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EMILIN1 Antibody (C11A8) - Azide and BSA Free, Novus Biologicals™
SDP

Rat Monoclonal Antibody

$243.50 - $560.50

Specifications

Antigen EMILIN1
Clone C11A8
Concentration LYOPH
Dilution Western Blot 2-10ug/ml, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
Applications Immunofluorescence, Immunohistochemistry (Frozen), Immunocytochemistry, Western Blot, Immunohistochemistry
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB019672
SDP
View Documents
Novus Biologicals
NBP2363380.02MG
0.02 mg
Each for $243.50
Only null left
Add to Cart
 
NBP236338
SDP
View Documents
Novus Biologicals
NBP236338
0.1 mg
Each for $560.50
Only null left
Add to Cart
 
Description

Description

EMILIN1 Monoclonal specifically detects EMILIN1 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.
Specifications

Specifications

EMILIN1
LYOPH
Immunofluorescence, Immunohistochemistry (Frozen), Immunocytochemistry, Western Blot, Immunohistochemistry
Unconjugated
Rat
Mouse
11117
recombinant mouse Emilin-1
Primary
Centrifuge vial prior to opening. Reconstitute in sterile water to a concentration of 0.1-1.0 mg/ml.
C11A8
Western Blot 2-10ug/ml, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
Monoclonal
Purified
RUO
DKFZp586M121, elastin microfibril interface-located protein 1, elastin microfibril interfacer 1, EMI, EMILIN, EMILIN-1, gp115
EMILIN1
IgG2a κ
Protein G purified
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.