Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EMILIN2 Antibody (828B3B3), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB355563 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB355563 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB355563 Supplier Novus Biologicals Supplier No. NBP236064MFV450
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

EMILIN2 Monoclonal antibody specifically detects EMILIN2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Frozen)

Specifications

Antigen EMILIN2
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone 828B3B3
Conjugate mFluor Violet 450 SE
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen
Formulation 50mM Sodium Borate
Gene Alias Elastin microfibril interface-located protein 2, elastin microfibril interfacer 2EMILIN-2, extracellular glycoprotein EMILIN-2, FLJ33200, FOAP-10, Protein FOAP-10
Host Species Mouse
Immunogen Recombinant gC1q domain of EMILIN2
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84034
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.