Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ EML 425
GREENER_CHOICE

Catalog No. p-7110859
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Reversible and non-competitive CBP/p300 inhibitor

EML 425 is a reversible and non-competitive CBP/p300 inhibitor (IC50 values are 1.1 and 2.9 μM, respectively). Cell permeable.

Specifications

Chemical Name or Material 5-[(4-Hydroxy-2,6-dimethylphenyl)methylene]-1,3-bis(phenylmethyl)-2,4,6(1H,3H,5H)-pyrimidinetrione
CAS 1675821-32-5
Quantity 10 mg
Target Histone Acetyltransferase Inhibitors
Molecular Formula C27H24N2O4
Formula Weight 440.49
Purity >98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.