Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EML1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$158.00 - $487.50
Specifications
Antigen | EML1 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP15659520
![]() |
Novus Biologicals
NBP15659520UL |
20 μL |
Each for $158.00
|
|
|||||
NBP156595
![]() |
Novus Biologicals
NBP156595 |
100 μL |
Each for $487.50
|
|
|||||
Description
EML1 Polyclonal specifically detects EML1 in Human samples. It is validated for Western Blot.Specifications
EML1 | |
Polyclonal | |
Rabbit | |
O00423 | |
2009 | |
Synthetic peptides corresponding to EML1(echinoderm microtubule associated protein like 1) The peptide sequence was selected from the C terminal of EML1. Peptide sequence YPCSQFRAPSHIYGGHSSHVTNVDFLCEDSHLISTGGKDTSIMQWRVI. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
echinoderm microtubule associated protein like 1, ELP79, EMAP1, EMAP-1, EMAPechinoderm microtubule-associated protein-like 1, EMAPL1, EMAPLhuEMAP-1, FLJ45033, HuEMAP, HuEMAP-1 | |
EML1 | |
IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title