Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EML2 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB096082 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB096082 20 μL
NB096081 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB096082 Supplier Novus Biologicals Supplier No. NBP30473520UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

EML2 Polyclonal specifically detects EML2 in Mouse samples. It is validated for Western Blot.

Specifications

Antigen EML2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias echinoderm microtubule associated protein like 2, echinoderm microtubule-associated protein-like 2, echinoderm MT-associaated protein (EMAP)-like protein 70, echinoderm MT-associated protein (EMAP)-like protein 70, ELP70, EMAP-2huEMAP-2, EMAP2microtubule-associated protein like echinoderm EMAP, EMAPL2, HuEMAP-2, TRTLT2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 150-415 of human EML2 (NP_036287.1).
Purification Method Affinity purified
Quantity 20 μL
Regulatory Status RUO
Research Discipline Vision
Primary or Secondary Primary
Gene ID (Entrez) 24139
Target Species Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.