Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EML3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP17959620UL
Description
EML3 Polyclonal specifically detects EML3 in Human samples. It is validated for Western Blot.Specifications
| EML3 | |
| Polyclonal | |
| Western Blot 1:1000 | |
| NP_694997 | |
| EML3 | |
| Synthetic peptide directed towards the N terminal of human EML3The immunogen for this antibody is EML3. Peptide sequence LLVRSGSTESRGGKDPLSSPGGPGSRRSNYNLEGISVKMFLRGRPITMYI. | |
| 20 μL | |
| Primary | |
| Human | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS and 2% Sucrose with 0.09% Sodium Azide | |
| echinoderm microtubule associated protein like 3 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 256364 | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction