Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EML4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA595554
Change view
Click to view available options
Quantity:
100 μg
Catalog No. Quantity
PIPA595554 100 μg
1 options

Catalog No. PIPA595554

Supplier: Invitrogen™ PA595554

Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: rat brain tissue, mouse brain tissue, mouse lung tissue. IHC: human appendicitis tissue, human gastric cancer tissue, human lung cancer tissue, human glioma tissue, human placenta tissue, human rectal cancer tissue, human sarcoma tissue, human tonsil tissue. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

Transcription of protein-coding genes can be reconstituted on naked DNA with only the general transcription factors and RNA polymerase II. However, this minimal system cannot transcribe DNA packaged into chromatin, indicating that accessory factors may facilitate access to DNA. One such factor, FACT, interacts specifically with histones H2A/H2B to effect nucleosome disassembly and transcription elongation. FACT is composed of an 80 kDa subunit and a 140 kDa subunit; this gene encodes the 140 kDa subunit.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EML4
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and 0.05mg sodium azide
Gene EML4
Gene Accession No. Q3UMY5, Q9HC35
Gene Alias 4930443C24Rik; AI644019; C2orf2; echinoderm microtubule associated protein like 4; echinoderm microtubule-associated protein-like 4; ELP120; EMAP-4; EMAPL4; EML4; OTTHUMP00000201447; OTTHUMP00000201455; Restrictedly overexpressed proliferation-associated protein; ropp 120; ROPP120
Gene Symbols EML4
Host Species Rabbit
Immunogen E.coli-derived human EML4 recombinant protein (Position: K207-N423).
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27436, 313861, 78798
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.