Learn More
Description
Specifications
Specifications
| Antigen | EMMPRIN/CD147 |
| Applications | ELISA, Immunofluorescence |
| Classification | Monoclonal |
| Clone | 41 |
| Conjugate | FITC |
| Dilution | ELISA, Immunocytochemistry/Immunofluorescence |
| Formulation | PBS |
| Gene Alias | 5F7, basigin, basigin (Ok blood group), CD147, CD147 antigen, Collagenase stimulatory factor, EMMPRINTCSF, Extracellular matrix metalloproteinase inducer, Leukocyte activation antigen M6, M6, OK, OK blood group antigen, Tumor cell-derived collagenase stimulatory factor |
| Host Species | Rabbit |
| Immunogen | This antibody was obtained from a rabbit immunized with purified, recombinant Mouse EMMPRIN/CD147 (NP_001070652.1; Met 1-Arg 209). |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
