Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EMMPRIN/CD147 Antibody (MEM-M6/2), DyLight 755, Novus Biologicals™
SDP

Catalog No. NB500561IR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB500561IR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB500561IR Supplier Novus Biologicals Supplier No. NB500561IR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

EMMPRIN/CD147 Monoclonal specifically detects EMMPRIN/CD147 in Human, Porcine samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen EMMPRIN/CD147
Applications Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone MEM-M6/2
Conjugate DyLight 755
Dilution Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin
Gene Alias 5F7, basigin, basigin (Ok blood group), CD147, CD147 antigen, Collagenase stimulatory factor, EMMPRINTCSF, Extracellular matrix metalloproteinase inducer, Leukocyte activation antigen M6, M6, OK, OK blood group antigen, Tumor cell-derived collagenase stimulatory factor
Gene Symbols BSG
Host Species Mouse
Immunogen A soluble recombinant form of CD147, CD147Rg, which consists of the cDNA coding for the entire extracellular region of CD147 fused to the DNA coding for the hinge region, CH2 and CH3 domain of human IgG1.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Signal Transduction, Vision
Primary or Secondary Primary
Gene ID (Entrez) 682
Test Specificity The antibody MEM-M6/2 recognizes extracellular Ig domain D1 of CD147 (Neurothelin), a 50-60 kDa type I transmembrane glycoprotein primarily expressed on all leukocytes, red blood cells, platelets and endothelial cells; it is not expressed by resting lymphocytes. The antibody MEM-M6/1 is a high-affinity antibody capable of binding to unstimulated peripheral blood T cells.
Target Species Human, Pig
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.