Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EMMPRIN/CD147 Antibody (OX-47), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. N1064848A70 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N1064848A70 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N1064848A70 Supplier Novus Biologicals Supplier No. NB10064848AF750
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

EMMPRIN/CD147 Monoclonal specifically detects EMMPRIN/CD147 in Rat samples. It is validated for CyTOF, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen EMMPRIN/CD147
Applications CyTOF, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OX-47
Conjugate Alexa Fluor 750
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias 5F7, basigin, basigin (Ok blood group), CD147, CD147 antigen, Collagenase stimulatory factor, EMMPRINTCSF, Extracellular matrix metalloproteinase inducer, Leukocyte activation antigen M6, M6, OK, OK blood group antigen, Tumor cell-derived collagenase stimulatory factor
Gene Symbols BSG
Host Species Mouse
Immunogen T blasts from a mixed lymphocyte reaction.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 682
Test Specificity NB100-64848 recognizes a membrane glycoprotein of MW 42-48 kD expressed at high levels on activated lymphocytes but lower levels on resting cells. The antigen is also present on some endothelia. The antigen contains two Ig-like domains and an unusual transmembrane domain with a charged residue. It is abundant on activated lymphocytes: it is the homologue of the chicken blood brain barrier antigen HT7.
Target Species Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.