Learn More
Description
Specifications
Specifications
| Antigen | EMMPRIN/CD147 |
| Applications | Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot |
| Classification | Monoclonal |
| Clone | OX114 |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:100-1:2000, Flow Cytometry 1:25-1:50, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 |
| Formulation | PBS (pH 7.4) with 0.09% Sodium Azide |
| Gene Alias | 5F7, basigin, basigin (Ok blood group), CD147, CD147 antigen, Collagenase stimulatory factor, EMMPRINTCSF, Extracellular matrix metalloproteinase inducer, Leukocyte activation antigen M6, M6, OK, OK blood group antigen, Tumor cell-derived collagenase stimulatory factor |
| Gene Symbols | BSG |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
