Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EMMPRIN/CD147 Antibody (OX114) - BSA Free, Novus Biologicals™

Rat Monoclonal Antibody
$206.50 - $499.50
Specifications
Antigen | EMMPRIN/CD147 |
---|---|
Clone | OX114 |
Concentration | 1.0 mg/mL |
Dilution | Western Blot 1:100-1:2000, Flow Cytometry 1:25-1:50, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 |
Applications | Immunohistochemistry (Frozen), Immunohistochemistry, Flow Cytometry, Western Blot |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017727
![]() |
Novus Biologicals
NB100655180.025MG |
0.025 mg |
Each for $206.50
|
|
|||||
NB10065518
![]() |
Novus Biologicals
NB10065518 |
0.125 mg |
Each for $499.50
|
|
|||||
Description
EMMPRIN/CD147 Monoclonal specifically detects EMMPRIN/CD147 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
EMMPRIN/CD147 | |
1.0 mg/mL | |
Immunohistochemistry (Frozen), Immunohistochemistry, Flow Cytometry, Western Blot | |
Unconjugated | |
Rat | |
Cancer, Signal Transduction, Vision | |
5F7, basigin, basigin (Ok blood group), CD147, CD147 antigen, Collagenase stimulatory factor, EMMPRINTCSF, Extracellular matrix metalloproteinase inducer, Leukocyte activation antigen M6, M6, OK, OK blood group antigen, Tumor cell-derived collagenase stimulatory factor | |
BSG | |
IgG1 | |
Protein G purified | |
NB100-65518 recognizes mouse CD147 (also known as basigin). CD147 is a cell surface glycoprotein that is widely expressed on leukocytes, red cells, platelets and endothelial cells. The reported functions of CD147 are diverse and include involvement in T-cell activation, cell adhesion and induction of matrix metalloproteinases. OX-114 recognizes both isoforms of CD147, the common form containing 2 Ig-like domains and the rare form containing 3 Ig-like domains. This antibody is routinely tested in flow cytometry on mouse splenocytes. Note that for staining of whole blood samples increased amounts of antibody may be required due to the presence of CD147 on erythrocytes. |
OX114 | |
Western Blot 1:100-1:2000, Flow Cytometry 1:25-1:50, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
Monoclonal | |
Purified | |
RUO | |
Mouse | |
682 | |
Fusion protein mCD147dO-2rCD4d3 +4 | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title