Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EMMPRIN/CD147 Antibody (OX114), DyLight 488, Novus Biologicals™
SDP

Catalog No. NB10065518G Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.125 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10065518G 0.125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10065518G Supplier Novus Biologicals Supplier No. NB10065518G
Add to Cart
Add to Cart

Rat Monoclonal Antibody

EMMPRIN/CD147 Monoclonal specifically detects EMMPRIN/CD147 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot.

Specifications

Antigen EMMPRIN/CD147
Applications Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot
Classification Monoclonal
Clone OX114
Conjugate DyLight 488
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias 5F7, basigin, basigin (Ok blood group), CD147, CD147 antigen, Collagenase stimulatory factor, EMMPRINTCSF, Extracellular matrix metalloproteinase inducer, Leukocyte activation antigen M6, M6, OK, OK blood group antigen, Tumor cell-derived collagenase stimulatory factor
Gene Symbols BSG
Host Species Rat
Immunogen Fusion protein mCD147dO-2rCD4d3 +4
Purification Method Protein G purified
Quantity 0.125 mL
Regulatory Status RUO
Research Discipline Cancer, Signal Transduction, Vision
Primary or Secondary Primary
Gene ID (Entrez) 682
Test Specificity NB100-65518 recognizes mouse CD147 (also known as basigin). CD147 is a cell surface glycoprotein that is widely expressed on leukocytes, red cells, platelets and endothelial cells. The reported functions of CD147 are diverse and include involvement in T-cell activation, cell adhesion and induction of matrix metalloproteinases. OX-114 recognizes both isoforms of CD147, the common form containing 2 Ig-like domains and the rare form containing 3 Ig-like domains. This antibody is routinely tested in flow cytometry on mouse splenocytes. Note that for staining of whole blood samples increased amounts of antibody may be required due to the presence of CD147 on erythrocytes.
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.