Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EMR1 Antibody, Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB353580 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB353580 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB353580 Supplier Novus Biologicals Supplier No. NBP306088JF549
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

EMR1 Polyclonal antibody specifically detects EMR1 in Human samples. It is validated for Western Blot

Specifications

Antigen EMR1
Applications Western Blot
Classification Polyclonal
Conjugate Janelia Fluor 549
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias ADGRE1, egf-like module containing, mucin-like, hormone receptor-like 1, egf-like module containing, mucin-like, hormone receptor-like sequence 1, EGF-like module receptor 1, EGF-like module-containing mucin-like hormone receptor-like 1, EMR1 hormone receptor, TM7LN3
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human EMR1.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, GPCR, Immune System Diseases, Immunology, Innate Immunity, Myeloid derived Suppressor Cell
Primary or Secondary Primary
Gene ID (Entrez) 2015
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.