Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EMR2 Antibody (494014), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NB303371 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB303371 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB303371 Supplier Novus Biologicals Supplier No. FAB48941X
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

EMR2 Monoclonal specifically detects EMR2 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen EMR2
Applications Flow Cytometry
Classification Monoclonal
Clone 494014
Conjugate Alexa Fluor 532
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias CD312, DKFZp781B135, egf-like module containing, mucin-like, hormone receptor-like 2, egf-like module containing, mucin-like, hormone receptor-like sequence 2, EGF-like module receptor 2, EGF-like module-containing mucin-like hormone receptor-like 2
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human EMR2, Gln24-Asn823, Accession # AAI27006
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 30817
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.