Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EMR3 Antibody (3D7), PE/Cy7, Novus Biologicals™
SDP

Catalog No. N1065935PC7 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N1065935PC7 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N1065935PC7 Supplier Novus Biologicals Supplier No. NB10065935PECY7
Add to Cart
Add to Cart

Armenian Hamster Monoclonal Antibody

EMR3 Monoclonal specifically detects EMR3 in Human samples. It is validated for Flow Cytometry, Immunoprecipitation.

Specifications

Antigen EMR3
Applications Flow Cytometry, Immunoprecipitation
Classification Monoclonal
Clone 3D7
Conjugate PE-Cyanine7
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias egf-like module containing, mucin-like, hormone receptor-like 3, EGF-like module receptor 3, EGF-like module-containing mucin-like hormone receptor-like 3, egf-like module-containing mucin-like receptor 3
Gene Symbols ADGRE3
Host Species Armenian Hamster
Immunogen ARHO-EMR3-CD97 (EGF1) transfectants.
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline GPCR, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 84658.0
Test Specificity NB100-65935 recognizes human epidermal growth factor (EGF) module-containing mucin-like hormone receptor 3 (EMR3), which is a 56kD member of the EGF-7 transmembrane (TM7) family of adhesion receptors. EMR3 is expressed at the cell surface as a heterodimer. The molecule is predominantly expressed on granulocytes, and at lower levels on mature myeloid cells, monocytes and dendritic cells. EMR3 is absent on lymphocytes, haematopoietic stem cells and myeloid progenitors. Studies suggest that the EMR3 molecule is up-regulated during late stages of neutrophil differentiation and is a marker for terminally differentiated cells. The exact functions of EMR3 and its ligands have not yet been determined.
Target Species Human
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.