Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Emx1 Antibody, mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB343945 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB343945 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB343945 Supplier Novus Biologicals Supplier No. NBP241151MFV610

Chicken Polyclonal Antibody

Emx1 Polyclonal antibody specifically detects Emx1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA

Specifications

Antigen Emx1
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias empty spiracles homeobox 1, empty spiracles-like protein 1
Host Species Chicken
Immunogen Antibody was raised against a 16 amino acid synthetic peptide near the amino terminus of human EMX1. The immunogen is located within amino acids 50 - 100 of EMX1. Amino Acid Squence: CAAAASEEPLRPTALN
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2016
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgY
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.