Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EMX1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA535373
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA535373 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA535373 Supplier Invitrogen™ Supplier No. PA535373
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

EMX1 Polyclonal Antibody for Western Blot

EMX1 is a transcription factor, which in cooperation with EMX2. It acts to generate the boundary between the roof and archipallium in the developing brain. It may function in combinations with OTX1/2 to specify cell fates in the developing central nervous system.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EMX1
Applications Western Blot
Classification Polyclonal
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene Emx1
Gene Accession No. Q04741, Q04742
Gene Alias empty spiracles homeobox 1; empty spiracles homolog 1; Empty spiracles-like protein 1; EMX1; Emx-1; Homeobox protein EMX1; RGD1564002
Gene Symbols Emx1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 228-257 amino acids from the C-terminal region of human EMX1.
Purification Method Ammonium sulfate precipitation
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13796, 2016
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.