Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EN-RAGE/S100A12 Antibody (133), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB109484 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB109484 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB109484 Supplier Novus Biologicals Supplier No. NBP289816AF405
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

EN-RAGE/S100A12 Monoclonal specifically detects EN-RAGE/S100A12 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen EN-RAGE/S100A12
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 133
Conjugate Alexa Fluor 405
Formulation 50 mM Sodium Borate
Gene Alias CAAF1Neutrophil S100 protein, CAGCS100 calcium binding protein A12 (calgranulin C), Calcium-binding protein in amniotic fluid 1, Calgranulin-C, CGRPEN-RAGE, ENRAGE, Extracellular newly identified RAGE-binding protein, MRP6, p6calgranulin C, protein S100-A12, S100 calcium binding protein A12, S100 calcium-binding protein A12, S100 calcium-binding protein A12 (calgranulin C)
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human EN-RAGE/S100A12 (NP_005612.1; Met1-Glu92).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cytokine Research, Immunology, Innate Immunity, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 6283
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.