Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EN-RAGE/S100A12 Antibody (OTI1D1), DyLight 680, Novus Biologicals™
SDP

Catalog No. NBP271276FR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP271276FR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP271276FR Supplier Novus Biologicals Supplier No. NBP271276FR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

EN-RAGE/S100A12 Monoclonal specifically detects EN-RAGE/S100A12 in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen EN-RAGE/S100A12
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI1D1
Conjugate DyLight 680
Dilution Western Blot, Immunohistochemistry
Formulation PBS with 0.05% Sodium Azide
Gene Alias CAAF1Neutrophil S100 protein, CAGCS100 calcium binding protein A12 (calgranulin C), Calcium-binding protein in amniotic fluid 1, Calgranulin-C, CGRPEN-RAGE, ENRAGE, Extracellular newly identified RAGE-binding protein, MRP6, p6calgranulin C, protein S100-A12, S100 calcium binding protein A12, S100 calcium-binding protein A12, S100 calcium-binding protein A12 (calgranulin C)
Gene Symbols S100A12
Host Species Mouse
Immunogen Full length recombinant protein of human EN-RAGE/S100A12 (NP_005612) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cancer, Cytokine Research, Immunology, Innate Immunity, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 6283.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.