Learn More
Pyramex™ Endeavor™ Plus Safety Eyewear
Lightweight and durable protective eyewear
$106.90 - $141.10
Specifications
Certifications/Compliance ANSI Z87.1,CSA Z94.3-07
Weight (Metric) 23.5 g
Lens Material Polycarbonate
View More Specs
Products 9
Catalog Number Mfr. No. Lens Coating Frame Color Lens Tint Price Quantity & Availability  
Catalog Number Mfr. No. Lens Coating Frame Color Lens Tint Price Quantity & Availability  
19-155-900
View Documents
Pyramex™
SBG9610S
Scratch-resistant Black, Gray Clear
Case of 12 for $110.60
Only null left
Add to Cart
19-155-901
View Documents
Pyramex™
SBG9610ST
Anti-fog, Scratch-resistant Black, Gray Clear
Case of 12 for $141.10
Only null left
Edge
Add to Cart
19-155-902
View Documents
Pyramex™
SBG9620S
Scratch-resistant Black, Gray Gray
Case of 12 for $119.50
Only null left
Add to Cart
19-155-903
View Documents
Pyramex™
SBG9620ST
Anti-fog, Scratch-resistant Black, Gray Gray
Case of 12 for $127.89
Only null left
Add to Cart
19-155-905
View Documents
Pyramex™
SBG9680S
Scratch-resistant Black, Gray Indoor/Outdoor, Mirror
Case of 12 for $126.90
Only null left
Add to Cart
19-155-906
View Documents
Pyramex™
SBGR9610S
Scratch-resistant Black, Green Clear
Case of 12 for $110.60
Only null left
Add to Cart
19-155-907
View Documents
Pyramex™
SBGR9620S
Scratch-resistant Black, Green Gray
Case of 12 for $119.50
Only null left
Add to Cart
19-155-908
View Documents
Pyramex™
SBGR9680S
Scratch-resistant Black, Green Indoor/Outdoor, Mirror
Case of 12 for $131.70
Only null left
Add to Cart
19-155-910
View Documents
Pyramex™
SBP9610S
Scratch-resistant Black, Pink Clear
Case of 12 for $106.90
Only null left
Add to Cart
Description
Description
  • Built-in brow guard shields against overhead particles
  • Special dielectric hinge design contains no metal parts
  • Curved, co-material temples allow for comfortable all day wear
  • Scratch resistant polycarbonate lens provides 99% UVA/B/C protection
  • 9.5 base one-piece wraparound lens provides excellent side protection
  • Soft and adjustable nose piece provides a customized fit to prevent slippping
  • Specifications
    Specifications
    ANSI Z87.1,CSA Z94.3-07
    23.5 g
    Polycarbonate
    Videos
    SDS
    Documents
    Documents
    Product Certifications
    Product Content Correction
    Your input is important to us. Please complete this form to provide feedback related to the content on this product.
    Product Title
    Your feedback has been submitted: Thank you for helping us improve our website.