Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Endocan/ESM-1 Antibody (RM0079-5J44) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NBP121710 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP121710 0.1 mg
NB018420 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP121710 Supplier Novus Biologicals Supplier No. NBP121710
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Endocan/ESM-1 Monoclonal specifically detects Endocan/ESM-1 in Mouse samples. It is validated for Western Blot.

Specifications

Antigen Endocan/ESM-1
Applications Western Blot
Classification Monoclonal
Clone RM0079-5J44
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:500-1:1000
Formulation Lyophilized from a 0.2 μm filtered solution in PBS. 0.025 mg size is provided in liquid form, PBS with No Preservative
Gene Alias endocan, endothelial cell-specific molecule 1, ESM-1
Gene Symbols ESM1
Host Species Rat
Immunogen Mouse recombinant Endocan
Purification Method Protein A or G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer
Primary or Secondary Primary
Gene ID (Entrez) 11082
Test Specificity This was produced from a hybridoma (mouse myeloma fused with spleen cells from a rat immunized with mouse recombinant Endocan.
Reconstitution Reconstitute with sterilized PBS to a final concentration of 0.5 mg/ml.
Target Species Mouse
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.