Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Endoglin/CD105 Antibody (166707), Allophycocyanin/Cy7, Novus Biologicals™
SDP

Catalog No. NB314936 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB314936 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB314936 Supplier Novus Biologicals Supplier No. FAB10971APCCY7
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Endoglin/CD105 Monoclonal specifically detects Endoglin/CD105 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen Endoglin/CD105
Applications Flow Cytometry
Classification Monoclonal
Clone 166707
Conjugate APC-Cyanine7
Dilution Flow Cytometry
Formulation PBS
Gene Alias CD105, CD105 antigen, endoglin, ENDOsler-Rendu-Weber syndrome 1, HHT1FLJ41744, ORW, ORW1
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Endoglin/CD105, Glu26-Gly586, Accession # Q5T9B9
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Cellular Markers, Endothelial Cell Markers, Extracellular Matrix, Hypoxia, Immunology, Mesenchymal Stem Cell Markers, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 2022
Target Species Human
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.