Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Endoglin/CD105 Antibody (ENG/1327), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP253334100UG
Description
Endoglin/CD105 Monoclonal specifically detects Endoglin/CD105 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifications
Endoglin/CD105 | |
Monoclonal | |
0.2 mg/mL | |
Western Blot 1 - 2 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, SDS-Page | |
CD105, CD105 antigen, endoglin, ENDOsler-Rendu-Weber syndrome 1, HHT1FLJ41744, ORW, ORW1 | |
Mouse | |
84 kDa | |
100 μg | |
Cancer, Cell Biology, Cellular Markers, Endothelial Cell Markers, Extracellular Matrix, Hypoxia, Immunology, Mesenchymal Stem Cell Markers, Stem Cell Markers | |
2022 | |
Store at 4C. | |
IgG1 |
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, SDS-Page | |
ENG/1327 | |
Unconjugated | |
10mM PBS with 0.05% BSA with 0.05% Sodium Azide | |
ENG | |
Recombinant human Endoglin protein fragment (aa74-251) (exact sequence is proprietary) | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction