Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Endoglin/CD105 Antibody (ENG/1327), Novus Biologicals™

Mouse Monoclonal Antibody
$268.00 - $524.00
Specifications
Antigen | Endoglin/CD105 |
---|---|
Clone | ENG/1327 |
Concentration | 0.2 mg/mL |
Dilution | Western Blot 1 - 2 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, SDS-Page |
Applications | Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, SDS-Page |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB25333420
![]() |
Novus Biologicals
NBP25333420UG |
20 μg |
Each for $268.00
|
|
|||||
NB253334100
![]() |
Novus Biologicals
NBP253334100UG |
100 μg |
Each for $524.00
|
|
|||||
NB253334200
![]() |
Novus Biologicals
NBP253334200UG |
200 μg | N/A | N/A | N/A | ||||
Description
Endoglin/CD105 Monoclonal specifically detects Endoglin/CD105 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifications
Endoglin/CD105 | |
0.2 mg/mL | |
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, SDS-Page | |
Unconjugated | |
Mouse | |
Cancer, Cell Biology, Cellular Markers, Endothelial Cell Markers, Extracellular Matrix, Hypoxia, Immunology, Mesenchymal Stem Cell Markers, Stem Cell Markers | |
10mM PBS with 0.05% BSA with 0.05% Sodium Azide | |
2022 | |
Recombinant human Endoglin protein fragment (aa74-251) (exact sequence is proprietary) | |
Primary | |
Store at 4C. |
ENG/1327 | |
Western Blot 1 - 2 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, SDS-Page | |
Monoclonal | |
Purified | |
RUO | |
Human | |
CD105, CD105 antigen, endoglin, ENDOsler-Rendu-Weber syndrome 1, HHT1FLJ41744, ORW, ORW1 | |
ENG | |
IgG1 | |
Protein A or G purified | |
84 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title