Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Endoglin/CD105 Antibody (MEM-226), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB500452646 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB500452646 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB500452646 Supplier Novus Biologicals Supplier No. NB500452JF646
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Endoglin/CD105 Monoclonal specifically detects Endoglin/CD105 in Human, Rat samples. It is validated for Flow Cytometry.

Specifications

Antigen Endoglin/CD105
Applications Flow Cytometry
Classification Monoclonal
Clone MEM-226
Conjugate Janelia Fluor 646
Dilution Flow Cytometry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CD105, CD105 antigen, endoglin, ENDOsler-Rendu-Weber syndrome 1, HHT1FLJ41744, ORW, ORW1
Gene Symbols ENG
Host Species Mouse
Immunogen Recombinant full length protein (Human). Expressed in vaccinia virus containing CD105 cDNA.
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline Cancer, Cell Biology, Cellular Markers, Endothelial Cell Markers, Extracellular Matrix, Hypoxia, Immunology, Mesenchymal Stem Cell Markers, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 2022
Test Specificity The antibody MEM-226 reacts with CD105 (Endoglin), a 90 kDa type I homodimerizing membrane glycoprotein expressed on vascular endothelial cells (small and large vessels), activated monocytes and tissue macrophages, stromal cells of certain tissues including bone marrow, pre-B lymphocytes in fetal marrow and erythroid precursors in fetal and adult bone marrow; it is also present on syncytiotrophoblast on placenta throughout pregnancy.
Target Species Human, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.