Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Endoglin/CD105 Antibody (MJ7/18), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB339409 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB339409 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB339409 Supplier Novus Biologicals Supplier No. NB10077666AF750
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Endoglin/CD105 Monoclonal antibody specifically detects Endoglin/CD105 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Flow (Cell Surface)

Specifications

Antigen Endoglin/CD105
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunoprecipitation
Classification Monoclonal
Clone MJ7/18
Conjugate Alexa Fluor 750
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Flow (Cell Surface)
Formulation 50mM Sodium Borate
Gene Alias CD105, CD105 antigen, endoglin, ENDOsler-Rendu-Weber syndrome 1, HHT1FLJ41744, ORW, ORW1
Host Species Rat
Immunogen Inflamed mouse skin
Purification Method Protein G purified
Quantity 0.5 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Cellular Markers, Endothelial Cell Markers, Extracellular Matrix, Hypoxia, Immunology, Mesenchymal Stem Cell Markers, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 2022
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.