Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Endoglin/CD105 Antibody (MJ7/18), PerCP, Novus Biologicals™
SDP

Catalog No. NB015712 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB015712 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB015712 Supplier Novus Biologicals Supplier No. NBP281085PCP
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Endoglin/CD105 Monoclonal specifically detects Endoglin/CD105 in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen Endoglin/CD105
Applications Flow Cytometry
Classification Monoclonal
Clone MJ7/18
Conjugate PerCP
Dilution Flow Cytometry
Formulation PBS
Gene Alias CD105, CD105 antigen, endoglin, ENDOsler-Rendu-Weber syndrome 1, HHT1FLJ41744, ORW, ORW1
Gene Symbols ENG
Host Species Rabbit
Immunogen MJ7/18 was prepared by immunizing rats with inflamed mouse skin tissue. The antibody was selected immunohistologically for reactivity with endothelial cells.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Cellular Markers, Endothelial Cell Markers, Extracellular Matrix, Hypoxia, Immunology, Mesenchymal Stem Cell Markers, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 2022
Test Specificity MJ7/18 binds specifically to mouse Endoglin/CD105. Endoglin/CD105 is a homodimer expressed by vascular endothelial cells. Endoglin/CD105 functions in adhesion and embryonic angiogenesis. Endoglin/CD105 is a major homodimeric glycoprotein of vascular endothelium involved in cell ahesion through integrin/RGD binding and the regulation of angiogenesis. Also acts as TGF-beta coreceptor and is involved in the TGF-beta/BMP signaling cascade. Endoglin/CD105 is also required for GDF2/BMP9 signaling through SMAD1 in endothelial cells.
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.