Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Endoglin/CD105 Antibody (SN6), Allophycocyanin/Cy7, Novus Biologicals™
SDP

Catalog No. N1065601AP7 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N1065601AP7 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N1065601AP7 Supplier Novus Biologicals Supplier No. NB10065601APCCY7
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Endoglin/CD105 Monoclonal specifically detects Endoglin/CD105 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot.

Specifications

Antigen Endoglin/CD105
Applications Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone SN6
Conjugate APC-Cyanine7
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias CD105, CD105 antigen, endoglin, ENDOsler-Rendu-Weber syndrome 1, HHT1FLJ41744, ORW, ORW1
Gene Symbols ENG
Host Species Mouse
Immunogen Partially purified cell membrane antigens from fresh leukemia cells.
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Cancer, Cell Biology, Cellular Markers, Endothelial Cell Markers, Extracellular Matrix, Hypoxia, Immunology, Mesenchymal Stem Cell Markers, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 2022.0
Test Specificity Recognizes the human CD105 cell surface antigen, a glycoprotein homodimer of 95kD subunits. CD105 is also known as endoglin, and is expressed by endothelial cells, activated monocytes and some leukemia cells.
Target Species Human
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.