Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Endoglycan/PODXL2 Antibody (MM0258- 7R2), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB361139 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB361139 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB361139 Supplier Novus Biologicals Supplier No. NBP212273MFV610
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Endoglycan/PODXL2 Monoclonal antibody specifically detects Endoglycan/PODXL2 in Human samples. It is validated for Western Blot

Specifications

Antigen Endoglycan/PODXL2
Applications Western Blot
Classification Monoclonal
Clone MM0258- 7R2
Conjugate mFluor Violet 610 SE
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias endoglycan, PODLX2, podocalyxin-like 2, podocalyxin-like protein 2
Host Species Mouse
Immunogen Human recombinant Endoglycan/PODXL2 (extracellular domain)
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 50512
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.