Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Endophilin B1/Bif-1 Antibody (30A882.1.1) - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP224733SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP224733SS 0.025 mg
NBP224733 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP224733SS Supplier Novus Biologicals Supplier No. NBP224733SS
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 26 publications

Endophilin B1/Bif-1 Monoclonal specifically detects Endophilin B1/Bif-1 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.

Specifications

Antigen Endophilin B1/Bif-1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
Classification Monoclonal
Clone 30A882.1.1
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot, Simple Western 1:200, Immunohistochemistry 5ug/ml, Immunocytochemistry/Immunofluorescence reported in multiple pieces of scientific literature, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 5 ug/ml
Gene Accession No. Q9Y371
Gene Alias Bax-interacting factor 1, Bif-1dJ612B15.2, CGI-61, endophilin-B1, KIAA0491endophilin B1, SH3 domain-containing GRB2-like protein B1, SH3-containing protein SH3GLB1, SH3-domain GRB2-like endophilin B1, SH3-domain, GRB2-like, endophilin B1
Gene Symbols SH3GLB1
Host Species Mouse
Immunogen A full-length human Bif-1 recombinant protein was used as immunogen.
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Apoptosis, Mitochondrial Mediated Pathway
Primary or Secondary Primary
Gene ID (Entrez) 51100
Test Specificity This antibody is reactive to Human.
Target Species Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.