Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Endophilin B1/Bif-1 Antibody (30A882.1.1) - BSA Free, Novus Biologicals™
SDP

Mouse Monoclonal Antibody has been used in 26 publications

$181.72 - $494.86

Specifications

Antigen Endophilin B1/Bif-1
Clone 30A882.1.1
Concentration 1.0 mg/mL
Dilution Western Blot, Simple Western 1:200, Immunohistochemistry 5ug/ml, Immunocytochemistry/Immunofluorescence reported in multiple pieces of scientific literature, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 5 ug/ml
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP224733SS
SDP
View Documents
Novus Biologicals
NBP224733SS
0.025 mg
Each for $181.72
Only null left
Add to Cart
 
NBP224733
SDP
View Documents
Novus Biologicals
NBP224733
0.1 mg
Each for $494.86
Only null left
Add to Cart
 
Description

Description

Endophilin B1/Bif-1 Monoclonal specifically detects Endophilin B1/Bif-1 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications

Specifications

Endophilin B1/Bif-1
1.0 mg/mL
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
Unconjugated
Mouse
Apoptosis, Mitochondrial Mediated Pathway
Q9Y371
51100
A full-length human Bif-1 recombinant protein was used as immunogen.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
30A882.1.1
Western Blot, Simple Western 1:200, Immunohistochemistry 5ug/ml, Immunocytochemistry/Immunofluorescence reported in multiple pieces of scientific literature, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 5 ug/ml
Monoclonal
Purified
RUO
Human, Mouse
Bax-interacting factor 1, Bif-1dJ612B15.2, CGI-61, endophilin-B1, KIAA0491endophilin B1, SH3 domain-containing GRB2-like protein B1, SH3-containing protein SH3GLB1, SH3-domain GRB2-like endophilin B1, SH3-domain, GRB2-like, endophilin B1
SH3GLB1
IgG1 κ
Protein G purified
This antibody is reactive to Human.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.