Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Endosialin/CD248/TEM1 Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB317495 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB317495 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB317495 Supplier Novus Biologicals Supplier No. NBP299057APC
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Endosialin/CD248/TEM1 Polyclonal antibody specifically detects Endosialin/CD248/TEM1 in Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen Endosialin/CD248/TEM1
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate APC
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias 2610111G01Rik, CD164 sialomucin-like 1, CD164L1, CD248 antigen, CD248 antigen, endosialin, CD248 molecule, endosialin, endosialin, MGC119479, TEM1MGC119478, Tumor endothelial marker 1
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human Endosialin/CD248/TEM1
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 57124
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.