Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Endosialin/CD248/TEM1 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP177311 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP177311 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP177311 Supplier Novus Biologicals Supplier No. NBP177311
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Endosialin/CD248/TEM1 Polyclonal specifically detects Endosialin/CD248/TEM1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence.

Specifications

Antigen Endosialin/CD248/TEM1
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5-2 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 2.5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 2.5 μg/mL
Formulation PBS with 0.02% Sodium Azide
Gene Accession No. NP_065137
Gene Alias 2610111G01Rik, CD164 sialomucin-like 1, CD164L1, CD248 antigen, CD248 antigen, endosialin, CD248 molecule, endosialin, endosialin, MGC119479, TEM1MGC119478, Tumor endothelial marker 1
Gene Symbols CD248
Host Species Rabbit
Immunogen Antibody was raised against a peptide corresponding to 14 amino acids near the amino terminus of human TEM-1. The immunogen is located within amino acids 40-90 of TEM-1 Amino Acid Squence: ATPRTPEEAQRVDS
Molecular Weight of Antigen 81 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57124
Test Specificity Human TEM-1 has 2 isoforms, including isoform 1 (757aa, 81kD), and isoform 2 (433aa, 46kD). Mouse TEM-1 has one isoform (765aa, 82kD) and Rat TEM-1 also has one isoform (764aa, 82kD). NBP1-77311 can detect human isoform 1, mouse and rat TEM-1.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.