Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Endosialin/CD248/TEM1 Antibody (OTI4B8), DyLight 488, Novus Biologicals™
SDP

Catalog No. NBP272075G Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP272075G 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP272075G Supplier Novus Biologicals Supplier No. NBP272075G
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Endosialin/CD248/TEM1 Monoclonal specifically detects Endosialin/CD248/TEM1 in Human samples. It is validated for Western Blot.

Specifications

Antigen Endosialin/CD248/TEM1
Applications Western Blot
Classification Monoclonal
Clone OTI4B8
Conjugate DyLight 488
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias 2610111G01Rik, CD164 sialomucin-like 1, CD164L1, CD248 antigen, CD248 antigen, endosialin, CD248 molecule, endosialin, endosialin, MGC119479, TEM1MGC119478, Tumor endothelial marker 1
Gene Symbols CD248
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 20-300 of human Endosialin/CD248/TEM1 (NP_065137) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 57124.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.