Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Endostatin Antibody (RM0060-3H11), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB361136 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB361136 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB361136 Supplier Novus Biologicals Supplier No. NBP212274APC
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Endostatin Monoclonal antibody specifically detects Endostatin in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen Endostatin
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone RM0060-3H11
Conjugate APC
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias antiangiogenic agent, collagen alpha-1(XVIII) chain, collagen, type XVIII, alpha 1, FLJ27325, FLJ34914, human type XVIII collagen10endostatin, KNO, KNO1MGC74745, Knobloch syndrome, type 1, KS, multi-functional protein MFP
Host Species Rat
Immunogen Mouse recombinant Endostatin
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Biologically Active Proteins, Cancer, Extracellular Matrix, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 80781
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.