Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Endostatin Antibody (RM0060-3H11) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB019139 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB019139 0.025 mg
NBP212274 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB019139 Supplier Novus Biologicals Supplier No. NBP2122740.025MG
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Endostatin Monoclonal specifically detects Endostatin in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Endostatin
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone RM0060-3H11
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500
Formulation A 0.2 um filtered solution in PBS with No Preservative
Gene Accession No. AAA19787
Gene Alias antiangiogenic agent, collagen alpha-1(XVIII) chain, collagen, type XVIII, alpha 1, FLJ27325, FLJ34914, human type XVIII collagen10endostatin, KNO, KNO1MGC74745, Knobloch syndrome, type 1, KS, multi-functional protein MFP
Gene Symbols COL18A1
Host Species Rat
Immunogen Mouse Endostatin
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80781
Reconstitution Reconstitute with 0.5 ml sterilized PBS to a final concentration of 0.2 mg/ml.
Target Species Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.