Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Endothelial Cells Antibody (MIL11), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10064612
Description
Endothelial Cells Monoclonal specifically detects Endothelial Cells in Porcine samples. It is validated for Immunohistochemistry, Immunohistochemistry-Frozen, Immunofluorescence.Specifications
Endothelial Cells | |
Monoclonal | |
Unconjugated | |
Endothelium | |
Plastic adherent porcine peripheral blood lymphocytes | |
1 mL | |
Primary | |
Pig | |
Supernatant |
Immunohistochemistry, Immunohistochemistry (Frozen) | |
MIL11 | |
Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:100-1:500 | |
Mouse | |
Tissue culture supernatant | |
RUO | |
NB100-64612 recognizes an antigen expressed by most endothelial cells in the pig. Expression is strongest upon intestinal endothelial cells, and staining is also seen upon endothelial cells in the lung, colon, trachea, thymus, lymph nodes, kidney and skin. In blood vessels MIL11 stains venous endothelial cells but not arterial endothelial cells. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgE |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction