Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Endothelial Cells Antibody (MIL11), Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$207.00 - $499.50

Specifications

Antigen Endothelial Cells
Clone MIL11
Dilution Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:100-1:500
Applications Immunohistochemistry (Frozen), Immunohistochemistry
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB017621
SDP
View Documents
Novus Biologicals
NB100646120.25ML
0.25 mL
Each for $207.00
Only null left
Add to Cart
 
NB10064612
SDP
View Documents
Novus Biologicals
NB10064612
1 mL
Each for $499.50
Only null left
Add to Cart
 
Description

Description

Endothelial Cells Monoclonal specifically detects Endothelial Cells in Porcine samples. It is validated for Immunohistochemistry, Immunohistochemistry-Frozen, Immunofluorescence.
Specifications

Specifications

Endothelial Cells
Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:100-1:500
Monoclonal
Supernatant
RUO
Endothelium
IgE
Tissue culture supernatant
NB100-64612 recognizes an antigen expressed by most endothelial cells in the pig. Expression is strongest upon intestinal endothelial cells, and staining is also seen upon endothelial cells in the lung, colon, trachea, thymus, lymph nodes, kidney and skin. In blood vessels MIL11 stains venous endothelial cells but not arterial endothelial cells.
MIL11
Immunohistochemistry (Frozen), Immunohistochemistry
Unconjugated
Mouse
Pig
Plastic adherent porcine peripheral blood lymphocytes
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.