Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Endothelial Cells Antibody (MIL11), Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NB100646120.25ML

 View more versions of this product

Catalog No. NB017621


Only null left
Add to Cart

Description

Description

Endothelial Cells Monoclonal specifically detects Endothelial Cells in Porcine samples. It is validated for Immunohistochemistry, Immunohistochemistry-Frozen, Immunofluorescence.
Specifications

Specifications

Endothelial Cells
Monoclonal
Unconjugated
PBS with 0.1% Sodium Azide
Mouse
Tissue culture supernatant
RUO
NB100-64612 recognizes an antigen expressed by most endothelial cells in the pig. Expression is strongest upon intestinal endothelial cells, and staining is also seen upon endothelial cells in the lung, colon, trachea, thymus, lymph nodes, kidney and skin. In blood vessels MIL11 stains venous endothelial cells but not arterial endothelial cells.
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgE
Immunohistochemistry (Frozen), Immunohistochemistry
MIL11
Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:100-1:500
Endothelium
Plastic adherent porcine peripheral blood lymphocytes
0.25 mL
Primary
Pig
Supernatant
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only