Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Endothelial Lipase Antibody, Novus Biologicals™
SDP

Catalog No. NB400118SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB400118SS 0.025 mL
NB400118 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB400118SS Supplier Novus Biologicals Supplier No. NB400118SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 2 publications

Endothelial Lipase Polyclonal specifically detects Endothelial Lipase in Human, Mouse (Negative) samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Endothelial Lipase
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA 1:100-1:2000. Use reported in scientific literature (PMID 22383927), Immunocytochemistry/Immunofluorescence 1:100
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.01% Sodium Azide
Gene Accession No. Q9Y5X9
Gene Alias EC 3.1.1, EC 3.1.1.3, EDLendothelial lipase, ELlipoprotein lipase H, Endothelial cell-derived lipase, lipase, endothelial, PRO719
Gene Symbols LIPG
Host Species Rabbit
Immunogen An N-terminal synthetic peptide made to human Endothelial Lipase [UniProt# Q9Y5X9]
Purification Method Affinity Purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Autophagy, Cholesterol Metabolism, Endothelial Cell Markers, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 9388
Test Specificity Human. Does not appear to recognize mouse.
Target Species Human, Mouse (Negative)
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.