Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Endothelial Lipase Antibody, DyLight 405, Novus Biologicals™
SDP

Catalog No. NB400111V Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB400111V 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB400111V Supplier Novus Biologicals Supplier No. NB400111V
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Endothelial Lipase Polyclonal specifically detects Endothelial Lipase in Human, Mouse (Negative) samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.

Specifications

Antigen Endothelial Lipase
Applications Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen)
Classification Polyclonal
Conjugate DyLight 405
Dilution Western Blot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Accession No. Q9Y5X9
Gene Alias EC 3.1.1, EC 3.1.1.3, EDLendothelial lipase, ELlipoprotein lipase H, Endothelial cell-derived lipase, lipase, endothelial, PRO719
Gene Symbols LIPG
Host Species Rabbit
Immunogen A C-terminal synthetic peptide made to human Endothelial Lipase [UniProt# Q9Y5X9]
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Autophagy, Cholesterol Metabolism, Endothelial Cell Markers, Lipid and Metabolism, Unfolded Protein Response
Primary or Secondary Primary
Gene ID (Entrez) 9388
Test Specificity Human. Predicted to react with monkey based on 100% sequence homology. Does not appear to recognize mouse.
Target Species Human, Mouse (Negative)
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.