Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Endothelial Lipase Antibody, PE, Novus Biologicals™
SDP

Catalog No. NB338792 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB338792 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB338792 Supplier Novus Biologicals Supplier No. NB400118PE
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Endothelial Lipase Polyclonal antibody specifically detects Endothelial Lipase in Human, Mouse (Negative) samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen Endothelial Lipase
Applications Western Blot, ELISA, Immunocytochemistry
Classification Polyclonal
Conjugate PE
Dilution Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Formulation PBS
Gene Alias EC 3.1.1, EC 3.1.1.3, EDLendothelial lipase, ELlipoprotein lipase H, Endothelial cell-derived lipase, lipase, endothelial, PRO719
Host Species Rabbit
Immunogen An N-terminal synthetic peptide made to human Endothelial Lipase [UniProt# Q9Y5X9]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Autophagy, Cholesterol Metabolism, Endothelial Cell Markers, Lipid and Metabolism, Microautophagy, Unfolded Protein Response
Primary or Secondary Primary
Gene ID (Entrez) 9388
Target Species Human, Mouse (Negative)
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.