Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Endoxifen

Potent antiestrogen; ERα ligand

Specifications

Purity >98%
CAS 110025-28-0
Product Type Endoxifen
Molecular Weight (g/mol) 373.49
Inhibitors Endoxifen
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
37-051-0R
View Documents
Tocris Bioscience™
3705/10
10 mg N/A N/A N/A
37-055-0R
View Documents
Tocris Bioscience™
3705/50
50 mg N/A N/A N/A
Description

Description

Estrogen receptor α (ERα) ligand; potent antiestrogen. Metabolite of tamoxifen (Cat. No. 0999). Primary metabolite responsible for the effectiveness of tamoxifen in ER-positive breast cancer.
Specifications

Specifications

>98%
Endoxifen
Endoxifen
110025-28-0
373.49
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.