Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Endrin (For EPA 500, 600, 8000/ Appendix IX, CLP Methods)

Catalog No. uspp2301
Encompass
Click to view available options
Quantity:
1 mL (in Methanol)
This item is not returnable. View return policy

Catalog No. US-PP-2301

Supplier: Agilent Technologies PP2301

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Endrin at 100μg/mL in Methanol

Chemical Identifiers

CAS 72-20-8
Molecular Formula C12H8Cl6O
Molecular Weight (g/mol) 380.895
InChI Key DFBKLUNHFCTMDC-WXTGYIQRSA-N
PubChem CID 101260859
SMILES C1C2C3C(C1C4C2O4)C5(C(=C(C3(C5(Cl)Cl)Cl)Cl)Cl)Cl

Specifications

Packaging Ampule
Quantity 1 mL (in Methanol)
Chemical Name or Material Endrin
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.