Learn More
Vernier Engineering Projects with NI LabVIEW™ and Vernier Printed Lab Book
Shop All Vernier Software Products
Click to view available options
Grade:
College and High School
Description
Engineering Projects with NI LabVIEW™ and Vernier contains engaging hands-on projects for SensorDAQ, LabQuest, or LabQuest Mini. Students are introduced to engineering concepts and programming with NI LabVIEW software. An introductory knowledge of NI LabVIEW programming is assumed (see our lab book Hands-On Introduction to NI LabVIEW if your students are new to NI LabVIEW).This book introduces many important science and engineering concepts, including analog and digital input, sensors and how they are calibrated, feedback and control, analog and digital output, servo and stepper motors, PID control, pulse-width modulation, voltage dividers and Wheatstone bridges.NI LabVIEW software is sold separately by National Instruments.
Specifications
Specifications
| Includes | 12 ready-to-use student projects with challenge exercisesAccess to up-to-date versions of the experimentsEssential instructor information including construction and programming tips and extensionsWord-processing files of the student sections, so you can edit the files to match your teaching preferencesNI LabVIEW sample programs for all projects and challengesA generous site license—buy one book and duplicate the experiments for your class |
| Grade | College and High School |