Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

engrailed homeobox 2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP214947 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP214947 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP214947 Supplier Novus Biologicals Supplier No. NBP214947
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

engrailed homeobox 2 Polyclonal specifically detects engrailed homeobox 2 in Human samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen engrailed homeobox 2
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:5000-1:20000, Immunoprecipitation 1:100-1:500
Formulation PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal
Gene Accession No. P19622
Gene Alias AUTS10, engrailed homeobox 2, engrailed homolog 2, engrailed-2, Homeobox protein en-2, homeobox protein engrailed-2, hu-En-2
Gene Symbols EN2
Host Species Rabbit
Immunogen Full length human EN2 Recombinant protein.
Molecular Weight of Antigen 34 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2020
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.